Learn More
World Precision Instrument Metzenbaum Scissors, 14.5 cm, Straight
SDP
Catalog No. 50822608
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • Stainless steel
Catalog No. 50-822-608 Supplier World Precision Instrument Supplier No. 501252
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • Stainless steel