Learn More
Med Vet International Miltex Metzembaum Scissors, 7in., Curved, Delicate Pattern And Tungsten Carbide
Supplier:  Med Vet International MIL182ATC
Encompass_Preferred
The Miltex 182A-TC Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and Tungsten Carbide. They have a length of 7in..
Catalog No. 50-282-9092