Learn More
Med Vet International Miltex Metzenbaum Scissors, 7in, Straight
Catalog No. 502522293
Encompass_Preferred
The Miltex 5-181 Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and have a length of 7in..
Catalog No. 50-252-2293 Supplier Med Vet International Supplier No. MIL5181
The Miltex 5-181 Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and have a length of 7in..