Learn More
Med Vet International Miltex Metzenbaum Scissors, Supercut, Curved, Delicate, One Serrated Blade, German, 4.5in, 5-Sc-284
Catalog No. 502832167
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 4.5in in length. Curved serrated blades making them ideal for dissection and biopsy. Made with medical grade German steel.
Catalog No. 50-283-2167 Supplier Med Vet International Supplier No. MIL5SC284
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 4.5in in length. Curved serrated blades making them ideal for dissection and biopsy. Made with medical grade German steel.