Learn More
Med Vet International Miltex Metzenbaum Scissors, Curved, German, 5.5in, 50/Box, St5-180
Catalog No. 502830092
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Curved blades. 5.5in total length. German made steel.
Catalog No. 50-283-0092 Supplier Med Vet International Supplier No. MILST5180
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Curved blades. 5.5in total length. German made steel.