Learn More
Med Vet International Miltex Metzenbaum Scissors, Curved, Tungsten Carbide, German, 7in, Pm-2750
Catalog No. 502829137
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Curved blades. Kept in most operating rooms, surgical bays, and other medical settings. 7in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-282-9137 Supplier Med Vet International Supplier No. MILPM2750
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Curved blades. Kept in most operating rooms, surgical bays, and other medical settings. 7in total length. Made with high-quality, medical grade German steel.