Learn More
Med Vet International Miltex Metzenbaum Dissecting Scissors, Curved, Supercut, German, 7in, Pm-6813
Supplier: Med Vet International MILPM6813
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Curved SuperCut blades that cut cleanly every time. 7in total length. German made steel.