Learn More
Med Vet International Miltex Metzenbaum-Nelson Scissors, Standard Pattern, Curved, Blunt Points, German, 11.25in, 5-192
Catalog No. 502831825
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Curved blades with blunt points. Standard pattern. 11.25in total length. Made with high-quality German steel.
Catalog No. 50-283-1825 Supplier Med Vet International Supplier No. MIL5192
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Curved blades with blunt points. Standard pattern. 11.25in total length. Made with high-quality German steel.