Learn More
Med Vet International Miltex Metzenbaum Scissors, Tungsten Carbide, Straight, Sharp Points, German, 7in, 5-181B-Tc
Catalog No. 502829768
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Straight blades with sharp points. Made with high-quality, medical grade German steel.
Catalog No. 50-282-9768 Supplier Med Vet International Supplier No. MIL5181BTC
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Straight blades with sharp points. Made with high-quality, medical grade German steel.