Learn More
World Precision Instrument Metzenbaum Scissors, 12.5cm, SuperCut, Straight, German
SDP
Catalog No. 50822283
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-822-283 Supplier World Precision Instrument Supplier No. 14214G
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.