Learn More
World Precision Instrument Metzenbaum Scissors, 18cm, Curved, Tungsten Carbide, German
SDP
Catalog No. 50822776
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-822-776 Supplier World Precision Instrument Supplier No. 501745G
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.