Learn More
Med Vet International Miltex Metzenbaum Scissors, 5.5in., Curved
Catalog No. 502831237
Encompass_Preferred
The Miltex 5-180 Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and have a length of 5.5in..
Catalog No. 50-283-1237 Supplier Med Vet International Supplier No. MIL5180
The Miltex 5-180 Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and have a length of 5.5in..