Learn More
Med Vet International Miltex Metzembaum Scissors, 7in, Curved, Tungsten Carbide
Supplier:  Med Vet International MIL5182TC
Encompass_Preferred
The Miltex 5-182TC Metzenbaum Scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. These scissors are made of German grade stainless steel and Tungesten Carbide. They have a length of 7in..
Catalog No. 50-264-3523