Learn More
Fine Science Tools Metzenbaum Fino Scissors, Tungsten Carbide, 14.5 cm
SDP
Catalog No. NC9942724
Encompass
Tip Shape: Straight/ Tips: Ultra Fine Blunt-Blunt/ Cutting Edge: 31mm/ Alloy / Material: Tungsten Carbide/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide
Catalog No. NC9942724 Supplier Fine Science Tools Supplier No. 1450815
Tip Shape: Straight/ Tips: Ultra Fine Blunt-Blunt/ Cutting Edge: 31mm/ Alloy / Material: Tungsten Carbide/ Length: 14.5cm/ Serrated: No/ Feature: Tungsten Carbide