Learn More
World Precision Instrument Metzenbaum Scissors, 11.5cm, SuperCut, Straight
SDP
Catalog No. NC0482628
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. NC0482628 Supplier World Precision Instrument Supplier No. 503269
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.