Learn More
Med Vet International Miltex Mixter Scissors, Curved, Blunt, German, 6.25in, 5-114
Catalog No. 502831779
Encompass_Preferred
Ratcheted and ring hemostatic scissors that are widely used for obscured surgical sites. Delicate and serrated jaws assure firm grip and cutting. Curved blades with blunt tips for dissection purposes. 6.25in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-1779 Supplier Med Vet International Supplier No. MIL5114
Ratcheted and ring hemostatic scissors that are widely used for obscured surgical sites. Delicate and serrated jaws assure firm grip and cutting. Curved blades with blunt tips for dissection purposes. 6.25in total length. Made with high-quality, medical-grade German steel.