Learn More
Fine Science Tools Extra Fine Bonn Scissors, 8.5cm
SDP
Catalog No. NC9668791
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 13mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No. Our finest iris scissors, best suited for microdissection under high magnification.
Catalog No. NC9668791 Supplier Fine Science Tools Supplier No. 1408508
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 13mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No. Our finest iris scissors, best suited for microdissection under high magnification.