Learn More
Med Vet International Miltex New'S Meisterhand Suture Scissors, Angled On Flat, German, 5.62in, Mh9-98
Catalog No. 502830336
Encompass_Preferred
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. MeisterHand scissors offer superior quality. The blades are angled to the side to make certain procedures easier. 5.62in in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0336 Supplier Med Vet International Supplier No. MILMH998
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. MeisterHand scissors offer superior quality. The blades are angled to the side to make certain procedures easier. 5.62in in total length. Made with high-quality, medical-grade German steel.