Learn More
World Precision Instrument Noyes Scissors, 12 cm, Curved, Sharp/Sharp, 15 mm Blades
SDP
Catalog No. 50822581
Encompass
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions. These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons.
These scissors are 12 cm (4.7") long and come with sharp/sharp, sharp/blunt, or blunt/blunt tips. On a sharp/sharp pair of scissors, both blades have sharp, pointed tips. These are used for precise cutting of delicate tissues. Use sharp/sharp tips when you need clean, accurate cuts.
Catalog No. 50-822-581 Supplier World Precision Instrument Supplier No. 501236
Noyes Micro Scissors are a surgical instrument designed for ophthalmic surgery and are an ideal choice for microdissection or for delicate small animal surgeries. Named after their inventor, ophthalmologist Dr. Arthur Noyes, the Noyes Micro Scissors are characterized by their small size and fine tips. They let you make precise and controlled incisions. These surgical scissors are typically used for delicate tasks such as cutting fine structures, trimming tissue, or creating small openings. Noyes scissors are a type of spring scissors which can be used ambidextrously. For this reason, Noyes scissors are often preferred by left-handed surgeons.
These scissors are 12 cm (4.7") long and come with sharp/sharp, sharp/blunt, or blunt/blunt tips. On a sharp/sharp pair of scissors, both blades have sharp, pointed tips. These are used for precise cutting of delicate tissues. Use sharp/sharp tips when you need clean, accurate cuts.