Learn More
Med Vet International Miltex Noyes Iris Straight Scissors, Sharp/Sharp, German, 4.5in, 18-1510
Catalog No. 502829145
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight blades with sharp/blunt tips. 4.5in total length. Made with high-quality, high-end German steel.
Catalog No. 50-282-9145 Supplier Med Vet International Supplier No. MIL181510
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight blades with sharp/blunt tips. 4.5in total length. Made with high-quality, high-end German steel.