Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder & Scissors, Serrated Tungsten Carbide, German, 4.75in, Pm-2169
Catalog No. 502829131
Encompass_Preferred
A needle holder, also called needle driver, is a surgical instrument, similar to a hemostat, used by doctors and surgeons to hold a suturing needle for closing wounds during suturing and surgical procedures. Coated with tungsten carbide for increased durability. 4.75in total. Made with high-quality, medical-grade German steel.
Catalog No. 50-282-9131 Supplier Med Vet International Supplier No. MILPM2169
A needle holder, also called needle driver, is a surgical instrument, similar to a hemostat, used by doctors and surgeons to hold a suturing needle for closing wounds during suturing and surgical procedures. Coated with tungsten carbide for increased durability. 4.75in total. Made with high-quality, medical-grade German steel.