Learn More
World Precision Instrument Operating Scissors, 11.5cm, Sharp/Blunt, Straight, German
SDP
Supplier:  World Precision Instrument 501753G
Encompass
Ring-handled Operating Scissors are a pair of straight surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 11.5 cm length is a smaller size that can be used for a variety of procedures, including biological and medical research. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved.
Catalog No. 50-822-790