Learn More
Roboz Surgical SCISSORS OPERATING 5 1/2 IN ST
SDP
Catalog No. NC9511027
Encompass
These 5 1/2-inch operating scissors are designed specifically for precise tissue dissection. Featuring a straight blade profile and sharp-blunt tips, they incorporate specialized cutting surfaces to ensure clean, efficient performance during surgical procedures.
  • One blade features micro-serrations perpendicular to the cutting edge for enhanced grip
  • Secondary blade is honed to razor sharpness for precise cutting
  • Matte grey rings and black screw provide easy visual identification
  • Intended exclusively for cutting soft tissue
Catalog No. NC9511027 Supplier Roboz Surgical Supplier No. RS6812SC
These 5 1/2-inch operating scissors are designed specifically for precise tissue dissection. Featuring a straight blade profile and sharp-blunt tips, they incorporate specialized cutting surfaces to ensure clean, efficient performance during surgical procedures.
  • One blade features micro-serrations perpendicular to the cutting edge for enhanced grip
  • Secondary blade is honed to razor sharpness for precise cutting
  • Matte grey rings and black screw provide easy visual identification
  • Intended exclusively for cutting soft tissue