Learn More
Med Vet International Miltex Mayo Meisterhand Dissecting Scissors, Straight, Sharp/Blunt, German, 5.12in, Mh5-12
Catalog No. 502831239
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.12in in length. Straight blades with sharp/blunt tips. MeisterHand scissors guarantee superior quality. Sharp/Blunt points. Made with high-quality, medical grade German metal.
Catalog No. 50-283-1239 Supplier Med Vet International Supplier No. MILMH514
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.12in in length. Straight blades with sharp/blunt tips. MeisterHand scissors guarantee superior quality. Sharp/Blunt points. Made with high-quality, medical grade German metal.