Learn More
Med Vet International Miltex Operating Scissors, Straight, Sharp/Sharp, 5.5in, 12 Pack, Eg5-6
Catalog No. 502829777
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. 5.5in total length. Straight blades. Made with high-quality, medical grade German steel.
Catalog No. 50-282-9777 Supplier Med Vet International Supplier No. MILEG56
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. 5.5in total length. Straight blades. Made with high-quality, medical grade German steel.