Learn More
Med Vet International Miltex Operating Scissors, Standard Pattern, Curved, Blunt/Blunt, German, 5.5in, MH5-56
Catalog No. 503046207
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. MeisterHand scissors provide excellent craftsmanship. 5.5in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-304-6207 Supplier Med Vet International Supplier No. MILMH556
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. MeisterHand scissors provide excellent craftsmanship. 5.5in total length. Made with high-quality, medical grade German steel.