Learn More
Med Vet International Miltex Mayo MeisterHand Dissecting Scissors, Straight, Standard Beveled Blades, German, 5.62in, MH5-16
Catalog No. 503045714
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.62in in length. Straight standard beveled blades. MeisterHand scissors guarantee superior quality. Made with high-quality, medical grade German metal.
Catalog No. 50-304-5714 Supplier Med Vet International Supplier No. MILMH516
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.62in in length. Straight standard beveled blades. MeisterHand scissors guarantee superior quality. Made with high-quality, medical grade German metal.