Learn More
Med Vet International Miltex Padgett Scissors, Curved, Blunt/Blunt, German, 4.5in, Pm-4250
Catalog No. 502829300
Encompass_Preferred
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. Curved with blunt/blunt tips. 4.5in total length. Made with high-end, medical grade German steel.
Catalog No. 50-282-9300 Supplier Med Vet International Supplier No. MILPM4250
Add to Cart
Add to Cart
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. Curved with blunt/blunt tips. 4.5in total length. Made with high-end, medical grade German steel.