Learn More
Med Vet International Miltex Padgett Strabismus Scissors, Straight Tungsten Carbide, German, 4.5in, Pm-2701
Catalog No. 502829135
Encompass_Preferred
Originally produced for ophthalmic procedures, Strabismus scissors are now used in a variety of procedures where delicate maneuvering is required. Straight tungsten carbide blades for prolonged sustainability. 4.5in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-282-9135 Supplier Med Vet International Supplier No. MILPM2701
Originally produced for ophthalmic procedures, Strabismus scissors are now used in a variety of procedures where delicate maneuvering is required. Straight tungsten carbide blades for prolonged sustainability. 4.5in total length. Made with high-quality, medical grade German steel.