Learn More
Med Vet International Miltex Potts-Smith Scissors, Angled On Side 45 Degrees, 14mm Blades, German, 7.in, 5-249B
Catalog No. 502829769
Encompass_Preferred
Crafted for use in a wide range of cardiothoracic surgical procedures requiring vascular incision or cutting. Being equipped with blunt tips which make them not only ideal for cutting, but also probing. The jaws are angled on the side making it easy for multiple surgical approaches. The blades are angled on the side 45 degrees. 14mm blades. 7in total length. Made with German steel.
Catalog No. 50-282-9769 Supplier Med Vet International Supplier No. MIL5249B
Crafted for use in a wide range of cardiothoracic surgical procedures requiring vascular incision or cutting. Being equipped with blunt tips which make them not only ideal for cutting, but also probing. The jaws are angled on the side making it easy for multiple surgical approaches. The blades are angled on the side 45 degrees. 14mm blades. 7in total length. Made with German steel.