Learn More
Med Vet International Miltex Potts-Smith Scissors, Angled On Side 60 Degrees, 15.5mm Blades, German, 7in, 5-249C
Catalog No. 502829767
Encompass_Preferred
Crafted for use in a wide range of cardiothoracic surgical procedures requiring vascular incision or cutting. Being equipped with blunt tips which make them not only ideal for cutting, but also probing. The jaws are angled on the side making it easy for multiple surgical approaches. The blades are angled on the side 60 degrees. 15.5mm blades. 7in total length. Made with German steel.
Catalog No. 50-282-9767 Supplier Med Vet International Supplier No. MIL5249C
Crafted for use in a wide range of cardiothoracic surgical procedures requiring vascular incision or cutting. Being equipped with blunt tips which make them not only ideal for cutting, but also probing. The jaws are angled on the side making it easy for multiple surgical approaches. The blades are angled on the side 60 degrees. 15.5mm blades. 7in total length. Made with German steel.