Learn More
Fine Science Tools Fine Scissors, Stainless Steel, Sharply Angled Up, 10.5 cm
SDP
Catalog No. NC0820118
Encompass
Tip Shape: Angled Up/ Tips: Sharp-Sharp/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC0820118 Supplier Fine Science Tools Supplier No. 1403710
Add to Cart
Add to Cart
Tip Shape: Angled Up/ Tips: Sharp-Sharp/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No