Learn More
Med Vet International Miltex Sistrunk Operating Scissors, Slightly Curved, German, 5.5in, 5-154
Catalog No. 502831895
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. Slightly curved blades. 5.5in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1895 Supplier Med Vet International Supplier No. MIL5154
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. Slightly curved blades. 5.5in total length. Made with high-quality, medical grade German steel.