Learn More
Med Vet International Miltex Spencer Stitch Scissors, Delicate, German, 3.5in, Mh9-100
Catalog No. 502830357
Encompass_Preferred
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. 3.5in in length. Delicate finish. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0357 Supplier Med Vet International Supplier No. MILMH9100
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. 3.5in in length. Delicate finish. Made with high-quality, medical-grade German steel.