Learn More
Med Vet International Miltex Spencer Stitch Scissors, 3in.
Catalog No. 503046567
Encompass_Preferred
The Miltex 9-100 Spencer Stitch Scissors are made of German grade stainless steel and have a length of 3in. These surgical scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal.
Catalog No. 50-304-6567 Supplier Med Vet International Supplier No. MIL9100
The Miltex 9-100 Spencer Stitch Scissors are made of German grade stainless steel and have a length of 3in. These surgical scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal.