Learn More
World Precision Instrument Spring Scissors, 14cm, Curved
SDP
Supplier:  World Precision Instrument 15906
Encompass
Spring scissors are fine surgical instruments for making small, precision cuts. They are 14cm long, and fit comfortably into your hand. The longer spring scissors may fit more naturally in a larger hand. WPI offers spring scissors that range from 9.5 to 14cm in length. The spring mechanism that allows for smooth, precise cutting action, which reduces the risk of tissue damage or trauma. Choose either straight or curved for the 8mm blade. The curved surgical scissors are used in ophthalmology to make precise incisions in the cornea or delicate tissues. The curved tip gives greater visibility at the tip of the scissors. The straight blades are more often used in microdissection, microsurgery or plastic surgery. The spring handled surgical scissors are preferred by left-handed surgeons, because they can be used ambidextrously. This gives the left-handed surgeon a more natural cutting action, which means the surgeon has greater control of the tip and the cutting movement.
Catalog No. 50-822-324