Learn More
Med Vet International Miltex Metzenbaum Scissors, Straight, Delicate Pattern, German, 8in, 5-183
Catalog No. 503046708
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 8in in length. Straight blades with a delicate pattern. Made with medical grade German steel.
Catalog No. 50-304-6708 Supplier Med Vet International Supplier No. MIL5183
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 8in in length. Straight blades with a delicate pattern. Made with medical grade German steel.