Learn More
Med Vet International Miltex Stevens Tenotomy Supercut Straight Scissors, Blunt/Blunt, German, 4.5in, 18-Sc-1472
Catalog No. 502831816
Encompass_Preferred
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Straight blades ending in blunt/blunt tips. 4.5in in length. These scissors are made with high-quality, medical-grade German steel.
Catalog No. 50-283-1816 Supplier Med Vet International Supplier No. MIL18SC1472
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Straight blades ending in blunt/blunt tips. 4.5in in length. These scissors are made with high-quality, medical-grade German steel.