Learn More
Med Vet International Miltex Meisterhand Stevens Tenotomy Scissors, Curved, Short Blades, Blunt/Blunt, German, 4.12in, Mh18-1466
Catalog No. 502831497
Encompass_Preferred
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. MeisterHand scissors guarantee signature exceptional craftsmanship. The scissors are equipped with short, curved blades culminating in blunt/blunt points. 4.12in in length. These scissors are made with high-quality, medical-grade German steel.
Catalog No. 50-283-1497 Supplier Med Vet International Supplier No. MILMH181466
 Restricted Item
Add to Cart
Add to Cart
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. MeisterHand scissors guarantee signature exceptional craftsmanship. The scissors are equipped with short, curved blades culminating in blunt/blunt points. 4.12in in length. These scissors are made with high-quality, medical-grade German steel.