Learn More
Med Vet International Miltex Straight Stevens Tenotomy Scissors, Blunt/Blunt, German, 4.12in, 18-1462
Catalog No. 502829140
Encompass_Preferred
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Blunt/blunt tips. 4.12in in length. These scissors are made with high-quality, medical-grade German steel.
Catalog No. 50-282-9140 Supplier Med Vet International Supplier No. MIL181462
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Blunt/blunt tips. 4.12in in length. These scissors are made with high-quality, medical-grade German steel.