Learn More
Med Vet International Miltex Curved Stevens Tenotomy Scissors, Blunt/Blunt, German, 6in, 18-1456
Catalog No. 502830424
Encompass_Preferred
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Blunt/Blunt tips. 6in in length. These scissors are made with high-quality, medical-grade German steel.
Catalog No. 50-283-0424 Supplier Med Vet International Supplier No. MIL181456
Typically used in the optical field, tenotomy scissors are mainly used in delicate surgical procedures. They are especially used in ophthalmic and neurosurgery. The small and sharp blades allows for precision cutting of the sensitive and delicate tissues in the eye region and of the small nerves. Blunt/Blunt tips. 6in in length. These scissors are made with high-quality, medical-grade German steel.