Learn More
Med Vet International Miltex Tenotomy Scissors, 4.5in., Straight
Supplier:  Med Vet International MIL181472
Encompass_Preferred
The Miltex 18-1472 Stevens Tenotomy Scissors have a length of 4.5in. and are made of German Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a straight tip.
Catalog No. 50-282-9142