Learn More
Med Vet International Miltex Strabismus Scissors, Straight, German 4in, 5-312
Catalog No. 502831360
Encompass_Preferred
Originally produced for ophthalmic procedures, Strabismus scissors are now used in a variety of procedures where delicate maneuvering is required. Straight tungsten carbide blades for prolonged sustainability. 4in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1360 Supplier Med Vet International Supplier No. MIL5312
Originally produced for ophthalmic procedures, Strabismus scissors are now used in a variety of procedures where delicate maneuvering is required. Straight tungsten carbide blades for prolonged sustainability. 4in total length. Made with high-quality, medical grade German steel.