Learn More
World Precision Instrument Student Vannas Spring Scissors, 9cm, Straight, 500 micron Tip
SDP
Supplier:  World Precision Instrument 501777
Encompass
Our Vannas spring scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, Vannas spring scissors are distinguished by their delicate, thin, sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Like all spring scissors, they are perfect for ambidextrous use and are often preferred by left-handed people. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. Our titanium scissors are lightweight and non-magnetic.
Catalog No. 50-822-822