Learn More
World Precision Instrument Mayo Scissors, 17cm, Straight, SuperCut w/TC, German
SDP
Catalog No. 50822617
Encompass
SuperCut Tungsten Carbide Mayo Scissors, also called Mayo Dissecting Scissors, are surgical scissors made popular by the Mayo Clinic. They have ring handles and long, straight, thick blades with semi-blunt tips that are straight or curved. The surgical scissors are 17 cm (6.75") long. The length of the blade determines the reach of the scissors when you are dissecting. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo Scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal. 
Catalog No. 50-822-617 Supplier World Precision Instrument Supplier No. 501259G
SuperCut Tungsten Carbide Mayo Scissors, also called Mayo Dissecting Scissors, are surgical scissors made popular by the Mayo Clinic. They have ring handles and long, straight, thick blades with semi-blunt tips that are straight or curved. The surgical scissors are 17 cm (6.75") long. The length of the blade determines the reach of the scissors when you are dissecting. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo Scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal. 