Learn More
Med Vet International Miltex Littler Suture Scissor, 4.625in., With Suture Hole in Blades
Supplier:  Med Vet International MIL21536
Encompass_Preferred
The Miltex 21-536 Littler Stitch Scissor is made of German grade stainless steel and feature a suture hole in the blades. They measure 4.625in. and have a curved tip with 2 sharp blades.
Catalog No. 50-283-0082