Learn More
World Precision Instrument Tenotomy Scissors, 10cm, SuperCut, Curved, German
SDP
Catalog No. 50822319
Encompass
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces. These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. These SuperCut tenotomy scissors are popular, because one blade is super sharp and the other is serrated. This lets you grip the tissue and shear for a clean cut the first time.
The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries. These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.
Catalog No. 50-822-319 Supplier World Precision Instrument Supplier No. 14396G
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces. These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. These SuperCut tenotomy scissors are popular, because one blade is super sharp and the other is serrated. This lets you grip the tissue and shear for a clean cut the first time.
The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries. These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.