Learn More
Med Vet International Miltex Tenotomy Stevens Scissors, 4.5in., Curved
Catalog No. 502829534
Encompass_Preferred
The Miltex 18-1476 Stevens Tenotomy Scissors have a length of 4.5in. and are made of German Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a curved tip.
Catalog No. 50-282-9534 Supplier Med Vet International Supplier No. 181476
The Miltex 18-1476 Stevens Tenotomy Scissors have a length of 4.5in. and are made of German Grade stainless steel. This Stevens Tenotomy Scissors are used for delicate dissection and cutting and have small, sharp blades, and has a curved tip.