Learn More
Med Vet International Miltex Utility Scissors, Curved, Sharp/Blunt, One Serrated Blade, Heavy Pattern, German, 6.5in, 1704
Catalog No. 503047165
Encompass_Preferred
Scissors designed for use in medical, surgical, or laboratory settings. Made to be used in a number of applications, such as surgical, education, veterinary, and much more. Molded with curved blades ending with sharp/blunt tips. One of the blades has serrations to prevent slippage. 6.5in total length. Put together with high-quality, medical grade German steel.
Catalog No. 50-304-7165 Supplier Med Vet International Supplier No. MIL1704
Scissors designed for use in medical, surgical, or laboratory settings. Made to be used in a number of applications, such as surgical, education, veterinary, and much more. Molded with curved blades ending with sharp/blunt tips. One of the blades has serrations to prevent slippage. 6.5in total length. Put together with high-quality, medical grade German steel.