Learn More
Med Vet International Miltex Utility Bandage Scissors, 7.5in.
Supplier:  Med Vet International MIL51000
Encompass_Preferred
The Miltex 5-1000 Utility Bandage Scissors are made of German Grade stainless steel and have a length of 7.5in. These bandage scissors have a plastic handle, an angle blade, and a blunt/blunt tip.
Catalog No. 50-283-1787